Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butopyronoxyl 95.0+%, TCI America™
SDP

Catalog No. D055025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D055025G 25 g
D0550500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D055025G Supplier TCI America Supplier No. D055025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 532-34-3
Molecular Formula C12H18O4
Molecular Weight (g/mol) 226.272
MDL Number MFCD00006574
InChI Key OKIJSNGRQAOIGZ-UHFFFAOYSA-N
Synonym Butyl 3,4-Dihydro-2,2-dimethyl-4-oxo-2H-pyran-6-carboxylate, 3,4-Dihydro-2,2-dimethyl-4-oxo-2H-pyran-6-carboxylic Acid Butyl Ester, Indalone
PubChem CID 10760
ChEBI CHEBI:82262
IUPAC Name butyl 2,2-dimethyl-4-oxo-3H-pyran-6-carboxylate
SMILES CCCCOC(=O)C1=CC(=O)CC(O1)(C)C

Specifications

Color White-Yellow
Boiling Point 113°C
Quantity 25 g
Formula Weight 226.27
Percent Purity ≥95.0% (HPLC)
Physical Form Liquid
Chemical Name or Material Butopyronoxyl
RTECSNumber UP7000000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.