Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl Alcohol, NF, 99.5%, Spectrum™ Chemical
SDP

Catalog No. 18601147 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-147 1 L Amber Glass Bottle
18-601-145 500 mL Amber Glass Bottle
18-601-146 6 x 500 mL Amber Glass Bottle
18-601-148 4 L Amber Glass Bottle
18-601-149 6 x 1 L Amber Glass Bottle
18-601-150 4 x 4 L Amber Glass Bottle
18-601-152 200 L Drum
18-601-151 20 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18-601-147 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B11971LT
Add to Cart
Add to Cart

B1197, 71-36-3, C4H10O

Spectrum™ Chemical Butyl Alcohol, NF is primarily a solvent and vehicle in many pharmaceutical formulations. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

CAS Max % 100%
Quantity 1 L
Packaging Amber Glass Bottle
UN Number UN1120
Boiling Point 117°C
Percent Purity 99.5%
Grade NF
Chemical Name or Material Butyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.