Learn More
Butyl Alcohol, Reagent, ACS, 99.4%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Description
Spectrum™ Chemical Butyl Alcohol, Reagent, ACS is more commonly known as butanol. It is also widely used as a solvent in textile and chemical fabrication processes. It can be used as a paint thinner and base for perfumes. As an ACS grade reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 6 x 500 mL |
| Packaging | Amber Glass Bottle |
| Residue after Evaporation | 0.005% |
| Titratable Acid | 0.0008 meq/g |
| UN Number | UN1120 |
| Water | 0.1% |
| Boiling Point | 117°C |
| Percent Purity | 99.4% |
| Grade | Reagent |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.