Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl Benzenesulfonate 98.0+%, TCI America™
SDP

Catalog No. B003125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B003125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B003125ML Supplier TCI America Supplier No. B003125ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 80-44-4
Molecular Formula C10H14O3S
Molecular Weight (g/mol) 214.28
MDL Number MFCD00025036
InChI Key NIKBCKTWWPVAIC-UHFFFAOYSA-N
Synonym Benzenesulfonic Acid Butyl Ester
PubChem CID 6642
IUPAC Name butyl benzenesulfonate
SMILES CCCCOS(=O)(=O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 153°C
Quantity 25 mL
Formula Weight 214.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Benzenesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.