Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl Cyanoacetate 98.0+%, TCI America™
SDP

Catalog No. C0440500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0440500G 500 g
C044025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0440500G Supplier TCI America Supplier No. C0440500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5459-58-5
Molecular Formula C7H11NO2
Molecular Weight (g/mol) 141.17
MDL Number MFCD00001941
InChI Key DJACTCNGCHPGOI-UHFFFAOYSA-N
Synonym butyl cyanoacetate, n-butyl cyanoacetate, acetic acid, cyano-, butyl ester, unii-f9h6l20voh, cyanoacetic acid butyl ester, cyanoacetic acid n-butyl ester, acetic acid, 2-cyano-, butyl ester, f9h6l20voh, butyl 2-cyanoethanoate, acmc-209lho
PubChem CID 79571
IUPAC Name butyl 2-cyanoacetate
SMILES CCCCOC(=O)CC#N

Specifications

Color Yellow
Boiling Point 231°C
Quantity 500 g
Formula Weight 141.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Cyanoacetate
EINECSNumber (2)-1549
RTECSNumber AG3692500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.