Learn More
Sigma Organic Chemistry Butyl myristate | 10mg | 110-36-1 | MFCD00015077
Supplier: Sigma Organic Chemistry CDS00717510MG

Butyl myristate, 10mg
About this Item:
CAS #: 110-36-1
MDL #: MFCD00015077
Molecular Weight: 284.48
UNSPSC Code: 12352200
Chemical Formula: C18H36O2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.