Learn More
Sigma Organic Chemistry Butyl vinyl ether | 100ML | 111-34-2 | MFCD00009454 | 0.98
Supplier: Sigma Organic Chemistry 110299100ML

Butyl vinyl ether, 100ML
About This Item:
Synonym(s): (Butyloxy)ethylene; Vinyl butyl ether
Linear Formula: CH3(CH2)3OCH=CH2
Molecular Weight: 100.16
CAS Number: 111-34-2
MDL Number: MFCD00009454
UNSPSC Code: 12162002
Purity: 0.98
Boiling Point: 94 C (lit.)
Melting Point: -92 C (lit.)
Density: 0.774 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.400 (lit.)
Storage: 2-8C
EINECS Number: 203-860-7
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.