Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Butylated Hydroxy Toluene
SCI_ED

Catalog No. s25212a Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Granules
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25212A 100 g Granules
S25212 500 g Granules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25212A
Only null left
Add to Cart
Add to Cart

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 128-37-0
Molecular Formula C15H24O
Molecular Weight (g/mol) 220.356
InChI Key NLZUEZXRPGMBCV-UHFFFAOYSA-N
Synonym 2,6-di-tert-butyl-4-methylphenol, butylated hydroxytoluene, butylhydroxytoluene, 2,6-di-tert-butyl-p-cresol, 2,6-di-t-butyl-4-methylphenol, ionol, dbpc, dibunol, stavox, bht
PubChem CID 31404
ChEBI CHEBI:34247
IUPAC Name 2,6-ditert-butyl-4-methylphenol
SMILES CC1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C

Specifications

Color White
Density 1.048
Assay Percent Range 0.99
Packaging Granules
Linear Formula ((CH3)3C)2CH3C6H2OH
Quantity 100 g
Formula Weight 220.35
Chemical Name or Material Butylated Hydroxy Toluene
ShelfLife 2 years
DOTInformation DOT Class 9, III, : Miscellaneous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.