Learn More
Medchemexpress LLC Butyric acid-13C4 sodium | 312623-84-0 | 98.0% | 113.06 g/mol | C13C3H7NaO2 | 5mg
SDP
Supplier:  Medchemexpress LLC HYW7507965MG
Encompass_Preferred
Butyric acid-13C4 sodium (Butanoic acid-13C4 sodium) is a stable isotope labeled compound with the activity of promoting cell proliferation and regulating gene expression Butyric acid-13C4 sodium can be used in metabolic research and compound development to help scientists gain a deeper understanding of the role of short-chain fatty acids in organisms Butyric acid-13C4 sodium also plays an important role in nutrition and intestinal health research especially in the regulation of probiotic function and intestinal microbiota
Catalog No. 50-002-70421