Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Butyryl Cholinesterase (Horse)

Catalog No. 89937025 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 Units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-937-025 500 Units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-937-025 Supplier Abnova™ Supplier No. P5213
Add to Cart
Add to Cart

Used for Enzyme

#REF!

Specifications

For Use With (Application) Enzyme Activity
Molecular Weight (g/mol) 440kDa
Name Butyryl Cholinesterase(Horse)
Preparation Method Native protein purified from Horse Serum
Quantity 500 Units
Storage Requirements Store at 4°C.
Regulatory Status RUO
Common Name BUTYRYL
Assays Butyryl Cholinesterase Assay
Biological Activity ≥4 units per mg dry weight. One Unit hydrolyzes 1 ≥ mole of acetylchoine per minute at 25°C, pH 7.4.
Species Horse
Protein Tag None
Expression System Native protein purified from Horse Serum
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.