Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyrylthiocholine Iodide 99.0+%, TCI America™
SDP

Catalog No. B077525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B077525G 25 g
B07755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B077525G Supplier TCI America Supplier No. B077525G

Chemical Identifiers

CAS 1866-16-6
Molecular Formula C9H20INOS
Molecular Weight (g/mol) 317.229
MDL Number MFCD00011845
InChI Key WEQAAFZDJROSBF-UHFFFAOYSA-M
Synonym butyrylthiocholine iodide, s-butyrylthiocholine iodide, 2-butyrylthio-n,n,n-trimethylethanaminium iodide, s-n-butyrylthiocholine iodide, 2-mercaptoethyl trimethylammonium iodide butyrate, ethanaminium, n,n,n-trimethyl-2-1-oxobutyl thio-, iodide, propylcarbonylthioethyl trimethylammonium iodide, 1-2-trimethylamino ethylthio butan-1-one, iodide, ethanaminium, n,n,n-trimethyl-2-1-oxobutyl thio-, iodide 1:1
PubChem CID 74630
IUPAC Name 2-butanoylsulfanylethyl(trimethyl)azanium;iodide
SMILES CCCC(=O)SCC[N+](C)(C)C.[I-]

Specifications

Melting Point 171°C
Color White-Yellow
Quantity 25 g
Formula Weight 317.23
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Butyrylthiocholine Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.