Learn More
BD Cell Analysis 3P BUV496 Mouse Anti-Human CD56, Clone NCAM16.2 (also known as NCAM 16), 50 ug
Supplier:  BD Cell Analysis 3P 750479
Encompass
The NCAM16.2 monoclonal antibody specifically binds to human CD56. It recognizes an extracellular immunoglobulin-like domain common to 120, 140, and 180 kDa forms of CD56, also known as the neural cell adhesion molecule (NCAM), NKH1 or MSK39.
Catalog No. NC1713366
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.