Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BUV615 Human IgG1 (N297A), κ Isotype Control, BD OptiBuild™
Human Isotype Control
Supplier: BD Biosciences 756245
Description

Specifications
Flow Cytometry | |
X40.huIgG1N297A.rMAb (also known as X40.huIgG1 N297A.rMAb) | |
Brilliant Ultraviolet 615 | |
IgG1, kappa Isotype Control (Anti-KLH) | |
Keyhole limpet hemocyanin (KLH) | |
50 μg | |
Human | |
Liquid |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human | |
Affinity Chromatography | |
RUO | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction