Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUV661 Mouse Anti-Human IgA1, BD OptiBuild™

Catalog No. BDB772210 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB772210 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB772210 Supplier BD Biosciences Supplier No. 772210
Add to Cart
Add to Cart

Mouse Monoclonal Secondary Antibody

It is validated for Flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgA1
Applications Flow Cytometry
Classification Monoclonal
Clone B3506B4 (also known as DLDB4)
Concentration 0.2 mg/mL
Conjugate Brilliant Ultraviolet 661
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias IGHA1; Immunoglobulin alpha 1
Host Species Mouse
Immunogen Human IgA1 Myeloma Protein
Purification Method Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3493
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.