Learn More
Description
Specifications
Specifications
| Antigen | IL-10 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | JES3-9D7 |
| Conjugate | Brilliant Ultraviolet 737 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | CSIF; Cytokine synthesis inhibitory factor; TGIF |
| Host Species | Rat |
| Immunogen | Recombinant Human IL-10 |
| Purification Method | Affinity Chromatography |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.