Learn More
Pain Care Labs Buzzy™ XL Healthcare

Description
- Latex-free
- Intended for healthcare and multi-patient use.
- Includes 4 Universal Healthcare Wings, 4 Silicone Comfort Strap, 2 AAA Batteries (preinstalled in unit), 1 Instruction Manual
- Reusable medical device that should be thoroughly cleaned and reprocessed following your facility’s infection control protocol for non-critical equipment.
- Stand up to hospital grade cleansers.
- Use with disposable hospital tourniquets or press in place for pain relief on contact.
Specifications
Specifications
| Type | Medical Device |
| Color | Plain Black |
| For Use With (Application) | Needle Procedures |
| Material | Medical Grade Plastic |
| Size | X-Large |
| Dimensions (L x W x H) | 5.5 x 3 x 8.5 cm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.