Learn More
Biolegend Brilliant Violet 421 anti-human CD209 (DC-SIGN) Antibody, 100 tests
Supplier:  Biolegend 330118
Encompass_Preferred
CD209, known as Dendritic Cell-Specific Intercellular adhesion molecule 3 (ICAM-3)-Grabbing Nonintegrin (DC-SIGN), is a 44 kD type II transmembrane glycoprotein and a member of the C-type lectin family. CD209 is expressed on myeloid dendritic cells, placental macrophages, liver and placental endothelial cells.
Catalog No. NC1928196
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.