Learn More
Biolegend Brilliant Violet 421™ anti-mouse CD152
Supplier:  Biolegend 106311
Encompass_Preferred
Brilliant Violet 421™ anti-mouse CD152 [UC10-4B9]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 125 uL
Catalog No. 50-163-127
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.