Learn More
Biolegend Brilliant Violet 421™ anti-mouse CD34
Supplier:  Biolegend 119321
Encompass_Preferred
Brilliant Violet 421™ anti-mouse CD34 [MEC14.7]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, ICC; Size: 125 uL
Catalog No. 50-163-799