Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BV480 Mouse Anti-Human CD45RA

Catalog No. BDB566114 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566114 100 Tests
BDB566155 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB566114 Supplier BD Supplier No. 566114
Add to Cart
Add to Cart

CD45RA is expressed on approximately 40-50% of peripheral CD4+ T cells, 50% of peripheral CD8+ T cells and on a portion of B cells and monocytes. BD Horizon™ BV480 Mouse Anti-Human CD45RA antigen is expressed by naive and activated T cells. CD45RA-specific antibodies are useful for the study of the suppressor/inducer subpopulation of CD4+ lymphocytes.

  • With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
  • BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
TRUSTED_SUSTAINABILITY

Specifications

Product Type BV480 Mouse Anti-Human CD45RA
Quantity 100 Tests
For Use With (Application) Flow Cytometry
Cross Reactivity QC Testing: Human
Form BV480
Sample Volume 5 μL
Content And Storage Store undiluted at 4°C
Storage Buffer Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.