Learn More
Biolegend Brilliant Violet 650™ anti-human CD279 (PD-1)
Supplier:  Biolegend 367430
Encompass_Preferred
Brilliant Violet 650™ anti-human CD279 (PD-1) [NAT105]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-8351
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.