Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BW 723C86 hydrochloride, Tocris Bioscience™

Catalog No. 105950
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-595-0 50 mg
10-591-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 10-595-0 Supplier R&D Systems Supplier No. 1059/50
This item is not returnable. View return policy

5-HT2B agonist

Selective 5-HT2B receptor agonist.

Chemical Identifiers

CAS 160521-72-2
Molecular Formula C16H19ClN2OS
Molecular Weight (g/mol) 322.851
InChI Key PYJBJMIBANAOFJ-UHFFFAOYSA-N
Synonym bw 723c86 hydrochloride, 1-5-thiophen-2-ylmethoxy-1h-indol-3-yl propan-2-amine hydrochloride, alpha-methyl-5-2-thienylmethoxy-1h-indole-3-ethanamine monohydrochloride, bw 723c hplc, 1h-indole-3-ethanamine,a-methyl-5-2-thienylmethoxy-,hydrochloride 1:1
PubChem CID 5311036
IUPAC Name 1-[5-(thiophen-2-ylmethoxy)-1H-indol-3-yl]propan-2-amine;hydrochloride
SMILES CC(CC1=CNC2=C1C=C(C=C2)OCC3=CC=CS3)N.Cl

Specifications

Quantity 50 mg
Formula Weight 322.85
Percent Purity >99%
Chemical Name or Material BW 723C86 hydrochloride
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.