Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell BW™ Solo, Non-Wireless Single-Gas Detector

Catalog No. 19807574
Encompass_Preferred
Change view
Click to view available options
Components Measured:
Ammonia
Hydrogen
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
19-807-574 Hydrogen
19-807-573 Ammonia
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-807-574 Supplier Honeywell Supplier No. BWSRY

missing translation for 'validMainframeMsg'

Honeywell BW™ Solo, Non-Wireless Single-Gas Detector features BLE connectivity for real-time visibility into the status and safety of hazardous-area workers, allowing companies respond faster, more knowingly and decisively to safety incidents.

  • User-friendly operation
    • Easy-to-read display for multiple languages
      • Lightweight convenience
        • Ease of maintenance
          • Serviceability
            • The new 1-Series sensor for enhanced reliability
              • Connected Bluetooth Low Energy (BLE) capabilities
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Class I, Division 1, Groups A, B, C, D T4; Class I, Zone 0, AEx/Ex ia IIC T4 Ga -40°C ≤ Tamb ≤60°C; ATEX: Sira 19ATEX2004; X 0518 g II 1G Ex ia IIC T4 Ga -40°C ≤ Tamb ≤60°C;; g I M1 Ex ia I Ma -40°C ≤ Tamb ≤60°C;; IECEx: IECEx SIR 19.0004; Ex ia IIC T4 Ga / Ex ia I Ma -40°C ≤ Tamb ≤60°C;; FCC ID: SU3BWS1; IC: 20969-BWS1; RED: RE-D Directive 2014/53/EU
Detection Limits 0 to 1000 ppm
Components Measured Hydrogen
Height (Metric) 7 cm
Includes Detector with specified sensor, stainless-steel alligator clip and concussion-proof housing; Test Cap with 1 ft of hose; Quick Reference Guide
Length (Metric) 6.7 cm
Sensor Types Replacement hydrogen (H2) sensor (4-Series)
Weight (English) 3.6 to 4.1 oz.
Weight (Metric) 103 to 116 g
Width (Metric) 3.6 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.