Learn More
Honeywell™ BW™ Solo Single Gas Detector
Description
Honeywell BW™ Solo is a easy to service single-gas detector with BLE connectivity that provides real-time visibility into the status and safety of hazardous-area workers, helping companies respond faster, more knowingly and decisively to safety incidents.
- User-friendly operation
- Easy-to-read display for multiple languages
- Lightweight convenience
- Ease of maintenance
- Serviceability
- The new 1-Series sensor for enhanced reliability
- Connected Bluetooth Low Energy (BLE) capabilities
Specifications
Specifications
| Product Type | Single-Gas Detector with BLE Connectivity |
| Color | Black |
| Material | Lexan TPE Mixture and Plastic |
| For Use With (Application) | Carbon Monoxide |
| Height (Metric) | 7 cm |
| Includes | Detector complete with specified sensor, stainless-steel alligator clip and concussion-proof housing; Test Cap with 1 ft of hose; Quick Reference Guide |
| Length (Metric) | 3.6 cm |
| Weight (Metric) | 103 to 116 g |
| Width (Metric) | 6.7 cm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.