Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ BW™ Solo Single Gas Detector

Catalog No. 19817111
Encompass_Preferred
Change view
Click to view available options
For Use With (Application):
Carbon Monoxide
Hydrogen Sulfide
Color:
Black
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Color
19-817-111 Carbon Monoxide Yellow
19-817-113 Hydrogen Sulfide Yellow
19-817-110 Carbon Monoxide Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-817-111 Supplier Honeywell™ Supplier No. BWSMLY

Honeywell BW™ Solo is a easy to service single-gas detector with BLE connectivity that provides real-time visibility into the status and safety of hazardous-area workers, helping companies respond faster, more knowingly and decisively to safety incidents.

Honeywell BW™ Solo is a easy to service single-gas detector with BLE connectivity that provides real-time visibility into the status and safety of hazardous-area workers, helping companies respond faster, more knowingly and decisively to safety incidents.

  • User-friendly operation
  • Easy-to-read display for multiple languages
  • Lightweight convenience
  • Ease of maintenance
  • Serviceability
  • The new 1-Series sensor for enhanced reliability
  • Connected Bluetooth Low Energy (BLE) capabilities
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Material Lexan TPE Mixture and Plastic
For Use With (Application) Carbon Monoxide
Height (Metric) 7 cm
Includes Detector complete with specified sensor, stainless-steel alligator clip and concussion-proof housing; Test Cap with 1 ft of hose; Quick Reference Guide
Length (Metric) 3.6 cm
Weight (Metric) 103 to 116 g
Width (Metric) 6.7 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.