Learn More
Selleck Chemical LLC BX-795 10mg 702675-74-9
SDP
Supplier:  Selleck Chemical LLC S127410MG
Encompass_Preferred
BX-795 is a potent and specific PDK1 inhibitor with IC50 of 6 nM, 140- and 1600-fold more selective for PDK1 than PKA and PKC in cell-free assays, respectively. Meanwhile, in comparison to GSK3? more than 100-fold selectivity observed for PDK1. BX-795 modulates autophagy via inhibiting ULK1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1093
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.