Learn More
3M™ BX™ Eyewear
Catalog No. 19065401 Shop All 3M Products
Change view
Click to view available options
Frame Color:
Blue
Ocean Blue
Silver, Black
Lens Tint:
Clear
Gray
Indoor/Outdoor Mirror
Light Amber
Light Blue
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-065-401 Ocean Blue Indoor/Outdoor Mirror
19-130-2635 Silver, Black Gray
19-130-2634 Silver, Black Clear
19-065-407 Blue Gray
19-065-406 Blue Light Amber
19-065-405 Blue Light Blue
19-065-400 Ocean Blue Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Catalog No. 19-065-401 Supplier 3M™ Supplier No. 7000127503
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
With antifog lenses
  • High-impact polycarbonate lens absorbs 99.9% UV
  • Adjustable soft-tip temples feel comfortable even after extended wear
  • Soft nose bridge
  • Three lens angles for custom fit
  • 3M Catalog# 11380-00000-20, 11381-00000-20, 11471-00000-20, 11472-00000-20, 11523-00000-20, 11524-00000-20, 11525-00000-20
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3 standards.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI Z87.1-2015, CSA Z94.3-2010
Lens Tint Indoor/Outdoor Mirror
Frame Color Ocean Blue
No. per Case 20/Cs.
Model 11472-00000-20
Weight (Metric) 1.0267 kg