Learn More
3M™ BX™ Eyewear
With antifog lenses
Supplier:  3M™ 7000127514
 View more versions of this product
Catalog No. 19-065-405
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
This item is not returnable. View return policy
Description
Description
  • High-impact polycarbonate lens absorbs 99.9% UV
  • Adjustable soft-tip temples feel comfortable even after extended wear
  • Soft nose bridge
  • Three lens angles for custom fit
  • 3M Catalog# 11380-00000-20, 11381-00000-20, 11471-00000-20, 11472-00000-20, 11523-00000-20, 11524-00000-20, 11525-00000-20
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3 standards.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ANSI Z87.1-2015
Blue
Adjustable; soft-tip
Light Blue
11523-00000-20
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.