Description
- High-impact polycarbonate lens absorbs 99.9% UV
- Adjustable soft-tip temples feel comfortable even after extended wear
- Soft nose bridge
- Three lens angles for custom fit
- 3M Catalog# 11380-00000-20, 11381-00000-20, 11471-00000-20, 11472-00000-20, 11523-00000-20, 11524-00000-20, 11525-00000-20
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3 standards.
Specifications
Specifications
| Certifications/Compliance | ANSI Z87.1-2015 |
| Lens Tint | Light Amber |
| Frame Color | Blue |
| Model | 11524-00000-20 |
| Temple Style | Adjustable; soft-tip |