Learn More
Akoya Biosciences BX021—CY5-RX021
SDP
Supplier:  Akoya Biosciences 5350002
Encompass
BX021—Cy5-RX021 is used with the CODEX Conjugation kit for conjugation of your antibody of choice with CODEX Barcode BX021. Each kit contains one tube of the lyophilized ,BX021 Barcode for use in a single conjugation reaction, and two tubes of Cy5-RX021 Reporters for use in optimizing CODEX antibody titration and CODEX runs. There is sufficient volume for 50 µg antibody conjugation, which provides enough material for 10 to 50 stains depending on yield and titer.
Catalog No. NC1751183
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.