Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BXDC2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988559
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8559 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8559 Supplier Bioss Supplier No. BS8436R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Required for biogenesis of the 60S ribosomal subunit.

Specifications

Antigen BXDC2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Brix1
Gene Accession No. Q8TDN6
Gene Alias 1110064N10Rik; BRIX; brix domain containing 2; Brix domain-containing protein 2; BRIX1; BRX1, biogenesis of ribosomes; BRX1, biogenesis of ribosomes, homolog; BRX1, biogenesis of ribosomes, homolog (S. cerevisiae); Bxdc2; C76935; RGD1308508; ribosome biogenesis protein Brix; Ribosome biogenesis protein BRX1 homolog
Gene Symbols Brix1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human BXDC2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55299
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.