Learn More
Accuflow Systems Saint-Gobain FEP Surface Protector, Aluminum Backing, 4 in, 36 yd
Supplier:  Accuflow Systems D9860011
Encompass_Preferred
Bytac surface protector is a bench liner composed of a Teflon FEP top film bonded to a support backing of aluminum foil with a pressure-sensitive acrylic adhesive. Supplied as a roll, the material provides a non-stick, chemically resistant surface that simplifies cleanup and protects laboratory benches, fume hoods, and other work surfaces.
  • 4 in width, 36 yd length roll.
  • FEP top film for non-stick, chemical-resistant surface.
  • Aluminum foil support backing with pressure-sensitive acrylic adhesive.
  • Roll form for easy application and trimming to fit surfaces.
  • Intended for bench and fume hood protection in laboratory and clean work areas.
Catalog No. NC3654344