Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Walter Products™ C-60 Buckminsterfullerene Molecular Model
SCI_ED

Catalog No. S139059 Shop All Walter Products Products
Change view
Click to view available options
Grade:
6+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139059 6+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139059 Supplier Walter Products™ Supplier No. MOLE1011
Add to Cart
Add to Cart

Molecular model visually displaying the soccer ball-like structure of Buckminsterfullerene.

Commonly known as a Buckyball, this Buckminsterfullerene molecular model offers a unique structure excellent for visual display.

  • Made of durable and colorful plastic
  • Molecular geometry and bond angles can be visibly seen in the structure, making it great for studying
  • Excellent molecular model kit for students studying chemistry in middle school or university
  • Has a cage-like fused-ring structure that resembles a soccer ball
  • Trending as a new health supplement that has anti-aging and longevity effects

Specifications

For Use With (Application) Chemistry
Grade 6+
Material Plastic
Product Type Molecular Model
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.