Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ C-clamp
SDPSCI_ED

Catalog No. S139556 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Size:
1 in. (Opening)
2 in. (Opening)
3 in.
4 in. (Opening)
5 in. (Opening)
6 in. (Opening)
Catalog No. Size
S139556 5 in. (Opening)
S01793 1 in. (Opening)
S139554 2 in. (Opening)
S81060 3 in.
S139555 4 in. (Opening)
S139557 6 in. (Opening)
6 options
This item is not returnable. View return policy

Catalog No. S139556

Supplier: United Scientific™ CLMPC5

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Strong and sturdy C-clamp designed for heavy duty usage.

  • Strong cast iron frames.
  • Screw-thread mechanism that provides a sturdy grip.
  • Available with screw-thread jaw opens up to 2, 3, 4, 5, or 6 in.

Specifications

Type C-Clamp
Material Cast Iron
Size 5 in. (Opening)
Color Black
Jaw Type Single Adjustment
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.