Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ C-DIM 12
GREENER_CHOICE

Catalog No. 582110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58-211-0 10 mg
58-215-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 58-211-0 Supplier Tocris Bioscience™ Supplier No. 5821/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Nurr1 activator; inhibits NF-κB-dependent gene expression

Nurr1 activator. Inhibits NF-κB and cytokine expression in microglial cells by stabilizing nuclear corepressor proteins, which reduces binding of p65 to inflammatory gene promoters. Neuroprotective in a mouse Parkinson's disease model. Orally bioavailable.

Chemical Identifiers

CAS 178946-89-9
Molecular Formula C23H17ClN2

Specifications

Quantity 10 mg
Solubility Soluble to 100mM in DMSO and to 100mM in ethanol
Formula Weight 356.85
Physical Form Solid
Purity >98%
Molecular Weight (g/mol) 356.85
Chemical Name or Material 3,3'-[(4-Chlorophenyl)methylene]bis[1H-indole]
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.