Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C/EBP beta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527244 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527244 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527244 Supplier Invitrogen™ Supplier No. PA527244
  • $620.00
    $489.65 / Each of 1

    Save $130.35

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Recommended positive controls: U87-MG. Predicted reactivity: Pig (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene belongs to the forkhead family of transcription factors which is characterized by a distinct DNA-binding forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of mesenchymal tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C/EBP beta
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene Cebpb
Gene Accession No. P17676
Gene Alias AGP/EBP; C EBP; c/EBP beta; C/EBPb; C/EBPbeta; C/EBP-beta; C/EBP-related protein 2; CCAAT enhancer binding protein beta; CCAAT/enhancer binding protein (C/EBP), beta; CCAAT/enhancer binding protein beta; CCAAT/enhancer-binding protein beta; CEBPB; Crp2; Il6dbp; IL-6DBP; interleukin 6-dependent DNA-binding protein; interleukin-6-dependent-binding protein; LAP; LIP; Liver activator protein; Liver-enriched inhibitory protein; Liver-enriched transcriptional activator; Nfil6; Nf-il6; NF-M; nuclear factor NF-IL6; nuclear factor of interleukin 6; nuclear factor-IL6; nuclear protein Il6; OTTMUSP00000017503; PP9092; Sfb; SF-B; Silencer factor B; TCF5; TCF-5; transcription factor 5; transcription factor C/EBP beta
Gene Symbols Cebpb
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human C/EBP beta. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1051
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.