Learn More
Description
Reconstitute in 500 μL of sterile water. Centrifuge to remove any insoluble material. After reconstitution keep aliquots at -20 °C for a higher stability, and at 4 °C with an appropriate antibacterial agent. Glycerol (1:1) may be added for an additional stability. Avoid repetitive freeze/thaw cycles.
Specifications
Specifications
| Antigen | c-Fos |
| Applications | Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | FOS |
| Gene Accession No. | P12841 |
| Gene Alias | activator protein 1; AP-1; cellular oncogene c-fos; cellular oncogene fos; cFos; C-FOS; c-fos oncogene; c-fos protooncogene protein; D12Rfj1; FBJ murine osteosarcoma viral (v-fos) oncogene homolog (oncogene FOS); FBJ murine osteosarcoma viral oncogene homolog; FBJ murine osteosarcoma viral oncogene homolog B; FBJ osteosarcoma oncogene; FBJ osteosarcoma oncogene B; FBJ osteosarcoma viral oncogene isoform deltaFosb-2; FOS; Fos proto-oncogene, AP-1 trancription factor subunit; Fos proto-oncogene, AP-1 transcription factor subunit; Fosb; FosB proto-oncogene, AP-1 transcription factor subunit; fra-2; G0/G1 switch regulatory protein 7; G0S7; I79_009428; p55; protein fosB; proto-oncogene cFOS; proto-oncogene c-Fos; proto-oncogene protein c-fos; v-fos FBJ murine osteosarcoma viral oncogene homolog |
| Gene Symbols | FOS |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
