Learn More
Bon Opus Biosciences Recombinant C. jejuni Cytolethal Distending Toxin B/CdtB (N-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP082500UG
Encompass_Preferred
Recombinant C. jejuni Cytolethal Distending Toxin B is produced by E.coli. The target gene encoding N27-F265 is expressed with a 6His tag at the N terminus. Expression system: E. coli. Accession# Q0PC57. Formulation: Supplied as a 0.2 um filtered solution of 2umM PB, 15umM NaCl, pH7.2. Bon Opus Cat# BP082
Catalog No. 50-208-0157
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.