Learn More
C.L. Sturkey EXTREMUS Disposable Microtome Blades
Catalog No. 22210035 Shop All CL Sturkey Products
Change view
Click to view available options
Profile:
High
Low
Quantity:
100 Pk.
120 Pk.
170 Pk.
200 Pk.
50 Pk.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Profile Quantity
22-210-035 Low 200 Pk.
22-210-034 High 100 Pk.
22-210-045 High 50 Pk.
22-210-048 Low 50 Pk.
22-210-036 Low 170 Pk.
22-210-033 High 120 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 22-210-035 Supplier C.L. Sturkey Supplier No. DT315XD
This item is not returnable. View return policy
Uniquely coated to suit the widest variety of cutting conditions
Offers a unique combination of durability and precision that saves time and money
Packaging Options
  • Standard Dispenser: Holds 50 blades per dispenser
  • Bulk Dispenser: Holds 120 Heavy Duty High-Profile or 200 Low-Profile blades per dispenser
  • Box Packaging: Holds 100 Heavy Duty High-Profile or 170 Low-Profile blades per box (non-dispensing)
Specifications
Profile Low
Disposable Single-use
Quantity 200 Pk.