Learn More
C.L. Sturkey EXTREMUS Disposable Microtome Blades
GSA_VA
Catalog No. 22210045 Shop All CL Sturkey Products
Change view
Click to view available options
Profile:
High
Low
Quantity:
100 Pk.
120 Pk.
170 Pk.
200 Pk.
50 Pk.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Profile Quantity
22-210-045 High 50 Pk.
22-210-034 High 100 Pk.
22-210-048 Low 50 Pk.
22-210-036 Low 170 Pk.
22-210-033 High 120 Pk.
22-210-035 Low 200 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 22-210-045 Supplier C.L. Sturkey Supplier No. D554X50
Add to Cart
Add to Cart
This item is not returnable. View return policy
Uniquely coated to suit the widest variety of cutting conditions
Offers a unique combination of durability and precision that saves time and money
Packaging Options
  • Standard Dispenser: Holds 50 blades per dispenser
  • Bulk Dispenser: Holds 120 Heavy Duty High-Profile or 200 Low-Profile blades per dispenser
  • Box Packaging: Holds 100 Heavy Duty High-Profile or 170 Low-Profile blades per box (non-dispensing)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Profile High
Disposable Single-use
Quantity 50 Pk.