Learn More
C.L. Sturkey Gold Disposable Microtome Blades
Catalog No. 22210009 Shop All CL Sturkey Products
Change view
Click to view available options
Profile:
High
Low
Quantity:
100 Pk.
170 Pk.
50 Pk.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Profile Quantity
22-210009 Low 170 Pk.
22-210013 High 100 Pk.
22-210-049 Low 50 Pk.
22-210-046 High 50 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22-210009 Supplier C.L. Sturkey Supplier No. DT315G
Add to Cart
Add to Cart
The ultimate blade for cutting hard tissue and bone
High-performance specialty blade, tough and durable
Packaging Options
  • Standard Dispenser: Holds 50 blades per dispenser
  • Bulk Dispenser: Holds 120 Heavy Duty High-Profile or 200 Low-Profile blades per dispenser
  • Box Packaging: Holds 100 Heavy Duty High-Profile or 170 Low-Profile blades per box (non-dispensing)
Specifications
Profile Low
Disposable Single-use
Quantity 170 Pk.
Coating Titanium Nitride