Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ c-MAF Monoclonal Antibody (sym0F1), Brilliant Violet™ 421, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 404985542
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-498-5542 100 Tests
40-498-5541 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 40-498-5542 Supplier Invitrogen™ Supplier No. 404985542
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

c-maf is the cellular counterpart of oncogenic v-maf that belongs to the family of basic region leucine zipper domain transcription factors. The leucine-zipper domain is involved in the interaction with LRPICD. There are two forms of human c-maf mRNA, c-maf-long and c-maf-short. It is identified in the genome of the acute transforming avian retrovirus AS42. c-maf targets are IL-4 in Th2 cells, the crystalline genes in lens fiber cells, insulin gene in islet, p53 and L7 where it exerts its transcriptional role through binding to a Maf recognition element (MARE). It regulates Th2 differentiation and lineage-specific hematopoiesis. c-maf is a transcription factor for IL-10 gene expression in LPS-activated macrophages. Chromosomal aberration involving maf is found in some forms of multiple myeloma. It is expressed in myeloma cell lines and resting monocytes/macrophages.
TRUSTED_SUSTAINABILITY

Specifications

Antigen c-MAF
Applications Flow Cytometry
Classification Monoclonal
Clone sym0F1
Concentration 5 μL/Test
Conjugate Brilliant Violet 421
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene MAF
Gene Accession No. O75444, P54843
Gene Alias 2810401A20Rik; A230108G15Rik; Avian musculoaponeurotic fibrosarcoma (MAF) protooncogene; avian musculoaponeurotic fibrosarcoma (v-maf) AS42 oncogene homolog; AW047063; AYGRP; basic domain leucine zipper transcription factor; CCA4; cMaf; C-Maf; c-Maf long form; c-maf proto-oncogene; CTRCT21; fl13d12; hMafA; LOW QUALITY PROTEIN: transcription factor Maf; LOW QUALITY PROTEIN: transcription factor MafA; maf; MAF bZIP transcription factor; MAF bZIP transcription factor A; maf protein; Maf2; MAFA; Mafa homolog; MGC71685; pancreatic beta-cell-specific transcriptional activator; Proto-oncogene c-Maf; RGD1562627; RIPE3b1; RIPE3b1 factor; T lymphocyte c-maf long form; transcription factor Maf; Transcription factor Maf-2; transcription factor MafA; transcription factor RIPE3b1; v-maf avian musculoaponeurotic fibrosarcoma oncogene A; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog A; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog a (paralog a); v-maf musculoaponeurotic fibrosarcoma oncogene family, protein A; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein A (avian); v-maf musculoaponeurotic fibrosarcoma oncogene homolog; V-maf musculoaponeurotic fibrosarcoma oncogene homolog A; wu:fl13d12; Z-cmaf
Gene Symbols MAF
Host Species Mouse
Immunogen Full-length E. coli-produced c-Maf.
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17132, 4094
Target Species Human, Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.