Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Maf Rabbit anti-Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501558044
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
50-155-8044 100 μL
1 options

Catalog No. 50-155-8044

Supplier: Bethyl Laboratories, Inc A300613A

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine

MAF acts as a transcriptional activator or repressor. The protein is involved in embryonic lens fiber cell development.

Specifications

Antigen c-Maf
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene Maf
Gene Accession No. P54843
Gene Alias Transcription factor Maf; Proto-oncogene c-maf; C-Maf; V-maf musculoaponeurotic fibrosarcoma oncogene homolog; MAF
Gene Symbols Maf
Host Species Rabbit
Immunogen Between 150 and 200
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17132
Target Species Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.