Learn More
CPC Scientific H-Glu-Val-Glu-Asp-Leu-Gln-Val-Arg-Asp-Val-Glu-Leu-Ala-Gly-Ala-Pro-Gly-Glu-Gly-Gly-Leu-Gln-Pro-Leu-Ala-Leu-Glu-Gly-Ala-Leu-Gln-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CPEP003B
Encompass
The C-peptide of this peptide is important for the biosynthesis of insulin, though for a long time has previously been considered to be biologically inert. Studies now point to C-peptide administration being accompanied by augmented blood flow in skeletal muscle and skin.ONE-LETTER SEQUENCE: EVEDLQVRDVELAGAPGEGGLQPLALEGALQMOLECULAR FORMULA: C137H225N37O49MOLECULAR WEIGHT:3174.5STORAGE CONDITIONS: -20 5CRESEARCH AREA: Diabetes
Catalog No. 50-210-9152
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.