Learn More
CPC Scientific H-Glu-Ala-Glu-Asp-Leu-Gln-Val-Gly-Gln-Val-Glu-Leu-Gly-Gly-Gly-Pro-Gly-Ala-Gly-Ser-Leu-Gln-Pro-Leu-Ala-Leu-Glu-Gly-Ser-Leu-Gln-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific CPEP001A
Encompass
This C-peptide measurement can sensitively assess beta-cell functions under standardized conditions, which is the most suitable primary outcome for clinical trials of therapies aimed at preserving or improving preserving endogenous insulin secretion in type 1 diabetes patients. ONE-LETTER SEQUENCE: EAEDLQVGQVELGGGPGAGSLQPLALEGSLQMOLECULAR FORMULA: C129H211N35O48MOLECULAR WEIGHT:3020.3STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [33017-11-7], RESEARCH AREA: DiabetesREFERENCES: A.S.C. Ko and D.G. Smyth, Eur. J. Biochem., 20, 190 (1971); P.E. Oyer et al., J. Biol. Chem., 246, 1375 (1971); K. Igano et al., Bull. Chem. Soc. Jpn., 54, 3088 (1981)
Catalog No. 50-210-9147
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.