Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C-Peptide, Mouse Anti-Human, Alexa Fluor™ 488, Clone: U8-424, BD Pharmingen™

Catalog No. BDB570993 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg;
Conjugate:
Alexa Fluor 488
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
BDB570993 0.1 mg Alexa Fluor 488
BDB571064 25 μg; Alexa Fluor 488
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB570993 Supplier BD Biosciences Supplier No. 570993
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C-Peptide Monoclonal antibody specifically detects C-Peptide in Human (QC Testing), Mouse,Rat (Tested in Development) samples. It is validated for Flow Cytometry (Intracellular Staining)
TRUSTED_SUSTAINABILITY

Specifications

Antigen C-Peptide
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone U8-424
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 488
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias INS; insulin; preproinsulin; proinsulin
Host Species Mouse
Immunogen Human C-Peptide
Purification Method The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3630, 16334, 24506
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.