Learn More
Enzo Life Sciences C-reactive protein (human), (recombinant) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT1640100
Encompass_Preferred
Alternative name: CRP, PTX1. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized from 0.22µm filtered solution in PBS. Generally 5-8% mannitol or trehalose is added as a protectant before lyophilization. Source: Produced in HEK 293 cells. Long Term Storage: -20°C.
Catalog No. 50-201-3952
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.