Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Rel Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501561084
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-1084 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-1084 Supplier Bethyl Laboratories, Inc Supplier No. A301825A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

The REL gene encodes c-Rel, a transcription factor that is a member of the Rel/NFKB family, which also includes RELA ., RELB (604758), NFKB1 ., and NFKB2 .. These proteins are related through a highly conserved N-terminal region termed the ′Rel domain, ′ which is responsible for DNA binding, dimerization, nuclear localization, and binding to the NFKB inhibitor . (Belguise and Sonenshein, 2007 [PubMed 18037997]).,

Specifications

Antigen c-Rel
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene REL
Gene Accession No. Q04864
Gene Alias C-Rel
Gene Symbols REL
Host Species Rabbit
Immunogen Between 325 and 375
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5966
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.