Learn More
ATCC Capnocytophaga sputigena Leadbetter et al. - ATCC 33612 (Type strain, BSL 2, research use only)
Supplier:  ATCC 33612
Encompass
Capnocytophaga sputigena Leadbetter et al. (ATCC 33612) is the genome-sequenced type strain isolated from a periodontal lesion. Supplied for laboratory research and quality-control applications, this BSL 2 organism requires anaerobic incubation on pre-reduced media and is intended for research use only.
  • Type strain and genome-sequenced.
  • Isolated from a periodontal lesion.
  • Biosafety level 2 organism.
  • Requires anaerobic atmosphere and incubation at 37 °C.
  • Grows best on pre-reduced peptone-yeast extract or trypticase soy media supplemented with glucose and blood.
  • Store refrigerated at 2 °C to 8 °C until use.
  • Intended for laboratory research and quality-control applications only; not for diagnostic, therapeutic, or consumption use.
Catalog No. NC3846327