Learn More
MBL International Corporation CycLex c-Src Kinase Assay/Inhibitor Screening Kit
SDP
Supplier:  MBL International Corporation CY1083
Encompass_Preferred
Size:96 Assays; Target:c-Src; The CycLex Research Product c-Src Kinase Assay/Inhibitor Screening Kit is designed to measure the activities of recombinant catalytic domain of c-Src for the rapid and sensitive evaluation of inhibitors or activators. It can be used for 96 assays.CycLex c-Src Kinase Assay/Inhibitor Screening Kit.
Catalog No. 50-455-34
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.